Merck
In vivo regulation of NGF-mediated functions by Nedd4-2 ubiquitination of TrkA.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2014-04-25)
Tao Yu, Laura Calvo, Begoña Anta, Saray López-Benito, Roger López-Bellido, Cristina Vicente-García, Lino Tessarollo, Raquel E Rodriguez, Juan C Arévalo
RÉSUMÉ
Trk neurotrophin receptor ubiquitination in response to ligand activation regulates signaling, trafficking, and degradation of the receptors. However, the in vivo consequences of Trk ubiquitination remain to be addressed. We have developed a mouse model with a mutation in the TrkA neurotrophin receptor (P782S) that results in reduced ubiquitination due to a lack of binding to the E3 ubiquitin ligase, Nedd4-2. In vivo analyses of TrkAP782S indicate that defective ubiquitination of the TrkA mutant results in an altered trafficking and degradation of the receptor that affects the survival of sensory neurons. The dorsal root ganglia from the TrkAP782S knock-in mice display an increased number of neurons expressing CGRP and substance P. Moreover, the mutant mice show enhanced sensitivity to thermal and inflammatory pain. Our results indicate that the ubiquitination of the TrkA neurotrophin receptor plays a critical role in NGF-mediated functions, such as neuronal survival and sensitivity to pain.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Nerve Growth Factor from Vipera lebetina venom, NGF, lyophilized powder, suitable for cell culture