Merck
Hypoxia after liver surgery imposes an aggressive cancer stem cell phenotype on residual tumor cells.
Annals of surgery (2013-11-21)
Klaas M Govaert, Benjamin L Emmink, Maarten W Nijkamp, Zing J Cheung, Ernst J A Steller, Szabolcs Fatrai, Menno T de Bruijn, Onno Kranenburg, Inne H M Borel Rinkes
RÉSUMÉ
To assess the contribution of hypoxia and bone marrow-derived cells to aggressive outgrowth of micrometastases after liver surgery. Liver surgery generates a microenvironment that fosters aggressive tumor recurrence. These areas are characterized by chronic hypoxia and influx of bone marrow-derived cells. The contribution of hematopoietic cell types was studied in mice lacking specific components of the immune system and in irradiated mice lacking all bone marrow-derived cells. Tumor cells were derived from colorectal cancer patients and from a metastatic tumor cell line. Hypoxia-induced changes in stem cell and differentiation marker expression, clone-forming potential, and metastatic capacity were assessed. The effect of vascular clamping on cancer stem cell (CSC) characteristics was performed in mice bearing patient-derived liver metastases. Immune cells and bone marrow-derived cells were not required for aggressive outgrowth of micrometastases in livers treated with surgery. Rather, hypoxia was sufficient to promote invasion and accelerate metastatic outgrowth. This was associated with a rapid loss of differentiation markers and increased expression of CSC markers and clone-forming capacity. Likewise, metastases residing in ischemia-reperfusion-injured liver lobes acquired CSC characteristics. Despite their renowned general resistance to chemotherapy, clone-forming CSCs were readily killed by the hypoxia-activated prodrug tirapazamine. Surgery-generated hypoxia in the liver causes rapid dedifferentiation of tumor cells into immature CSCs with high clone- and metastasis-forming capacity. The results help explain the phenomenon of aggressive local tumor recurrence after liver surgery and offer a potential strategy to kill aggressive CSCs by hypoxia-activated prodrugs.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Tirapazamine, ≥98% (HPLC)