Merck
Suppression of eIF2α kinases alleviates Alzheimer's disease-related plasticity and memory deficits.
Nature neuroscience (2013-08-13)
Tao Ma, Mimi A Trinh, Alyse J Wexler, Clarisse Bourbon, Evelina Gatti, Philippe Pierre, Douglas R Cavener, Eric Klann
RÉSUMÉ
Expression of long-lasting synaptic plasticity and long-term memory requires protein synthesis, which can be repressed by phosphorylation of eukaryotic initiation factor 2 α-subunit (eIF2α). Elevated phosphorylation of eIF2α has been observed in the brains of Alzheimer's disease patients and Alzheimer's disease model mice. Therefore, we tested whether suppressing eIF2α kinases could alleviate synaptic plasticity and memory deficits in Alzheimer's disease model mice. Genetic deletion of eIF2α kinase PERK prevented enhanced phosphorylation of eIF2α and deficits in protein synthesis, synaptic plasticity and spatial memory in mice that express familial Alzheimer's disease-related mutations in APP and PSEN1. Similarly, deletion of another eIF2α kinase, GCN2, prevented impairments of synaptic plasticity and defects in spatial memory exhibited by the Alzheimer's disease model mice. Our findings implicate aberrant eIF2α phosphorylation as a previously unidentified molecular mechanism underlying Alzheimer's disease-related synaptic pathophysioloy and memory dysfunction and suggest that PERK and GCN2 are potential therapeutic targets for treatment of individuals with Alzheimer's disease.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Anisomycin from Streptomyces griseolus, ≥98% (HPLC), solid
Sigma-Aldrich
Anisomycin, Ready Made Solution from Streptomyces griseolus, 10 mg/mL in DMSO
Sigma-Aldrich
Anti-CaM Kinase II Antibody, from rabbit, purified by affinity chromatography