Merck
  • Medical management to prevent recurrent nephrolithiasis in adults: a systematic review for an American College of Physicians Clinical Guideline.
Medical management to prevent recurrent nephrolithiasis in adults: a systematic review for an American College of Physicians Clinical Guideline.
Annals of internal medicine (2013-04-03)
Howard A Fink, Timothy J Wilt, Keith E Eidman, Pranav S Garimella, Roderick MacDonald, Indulis R Rutks, Michelle Brasure, Robert L Kane, Jeannine Ouellette, Manoj Monga
RÉSUMÉ
Optimum management to prevent recurrent kidney stones is uncertain. To evaluate the benefits and harms of interventions to prevent recurrent kidney stones. MEDLINE, Cochrane, and other databases through September 2012 and reference lists of systematic reviews and randomized, controlled trials (RCTs). 28 English-language RCTs that studied treatments to prevent recurrent kidney stones and reported stone outcomes. One reviewer extracted data, a second checked accuracy, and 2 independently rated quality and graded strength of evidence. In patients with 1 past calcium stone, low-strength evidence showed that increased fluid intake halved recurrent composite stone risk compared with no treatment (relative risk [RR], 0.45 [95% CI, 0.24 to 0.84]). Low-strength evidence showed that reducing soft-drink consumption decreased recurrent symptomatic stone risk (RR, 0.83 [CI, 0.71 to 0.98]). In patients with multiple past calcium stones, most of whom were receiving increased fluid intake, moderate-strength evidence showed that thiazides (RR, 0.52 [CI, 0.39 to 0.69]), citrates (RR, 0.25 [CI, 0.14 to 0.44]), and allopurinol (RR, 0.59 [CI, 0.42 to 0.84]) each further reduced composite stone recurrence risk compared with placebo or control, although the benefit from allopurinol seemed limited to patients with baseline hyperuricemia or hyperuricosuria. Other baseline biochemistry measures did not allow prediction of treatment efficacy. Low-strength evidence showed that neither citrate nor allopurinol combined with thiazide was superior to thiazide alone. There were few withdrawals among patients with increased fluid intake, many among those with other dietary interventions and more among those who received thiazide and citrate than among control patients. Reporting of adverse events was poor. Most trial participants had idiopathic calcium stones. Nearly all studies reported a composite (including asymptomatic) stone recurrence outcome. In patients with 1 past calcium stone, increased fluid intake reduced recurrence risk. In patients with multiple past calcium stones, addition of thiazide, citrate, or allopurinol further reduced risk. Agency for Healthcare Research and Quality.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Allopurinol, xanthine oxidase inhibitor
Sigma-Aldrich
Acetohydroxamic acid, 98%
Allopurinol, European Pharmacopoeia (EP) Reference Standard