Merck
A new tetracyclic triterpenoid compound from Mastich.
Journal of Asian natural products research (2013-03-09)
Jian-Bo Gao, Guo-Yu Li, Jian Huang, Hang-Yu Wang, Ke Zhang, Jin-Hui Wang
RÉSUMÉ
A new tetracyclic triterpenoid compound named as mastichinoic acid A (1) was isolated from Mastich. The structure was established as 3,7-dioxo-11β-hydroxy-5α-tirucalla-8,24(Z)-dien-26-oic acid by various spectroscopic techniques including MS, 1D, and 2D NMR and HRTOFMS.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Gum mastic