Merck
  • Solid state NMR of proteins at high MAS frequencies: symmetry-based mixing and simultaneous acquisition of chemical shift correlation spectra.
Solid state NMR of proteins at high MAS frequencies: symmetry-based mixing and simultaneous acquisition of chemical shift correlation spectra.
Journal of biomolecular NMR (2012-11-28)
Peter Bellstedt, Christian Herbst, Sabine Häfner, Jörg Leppert, Matthias Görlach, Ramadurai Ramachandran
RÉSUMÉ
We have carried out chemical shift correlation experiments with symmetry-based mixing sequences at high MAS frequencies and examined different strategies to simultaneously acquire 3D correlation spectra that are commonly required in the structural studies of proteins. The potential of numerically optimised symmetry-based mixing sequences and the simultaneous recording of chemical shift correlation spectra such as: 3D NCAC and 3D NHH with dual receivers, 3D NC'C and 3D C'NCA with sequential (13)C acquisitions, 3D NHH and 3D NC'H with sequential (1)H acquisitions and 3D CANH and 3D C'NH with broadband (13)C-(15)N mixing are demonstrated using microcrystalline samples of the β1 immunoglobulin binding domain of protein G (GB1) and the chicken α-spectrin SH3 domain.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Spectrin from human erythrocytes, buffered aqueous glycerol solution