Merck
  • Sulfur mustard cutaneous injury characterization based on SKH-1 mouse model: relevance of non-invasive methods in terms of wound healing process analyses.
Sulfur mustard cutaneous injury characterization based on SKH-1 mouse model: relevance of non-invasive methods in terms of wound healing process analyses.
Skin research and technology : official journal of International Society for Bioengineering and the Skin (ISBS) [and] International Society for Digital Imaging of Skin (ISDIS) [and] International Society for Skin Imaging (ISSI) (2012-06-30)
Cécile Cléry-Barraud, Nina Nguon, Virginie Vallet, Catherine Sentenac, Elise Four, Carine Arlaud, David Coulon, Isabelle Boudry
RÉSUMÉ
To date, sulphur mustard (SM) cutaneous toxicity has been commonly assessed on account of several animal models such as pigs and weanling pigs. Few experiments however, have been carried out on mice so far. In this study, we aimed at quantifying spontaneous wound healing processes after SM exposure on a SKH-1 mouse model through non-invasive methods over an extended period of time. Animals were exposed to 10 μL net SM in a vapor cup system. Measurements of barrier function (Transepidermal water loss), elasticity, skin color exposed to SM vapors were determined by evaporimetry, cutometer and image analysis on 23 animals up to 28 days. Results were subsequently correlated with histological and biochemical analyses. The TEWL parameter stands as a top-ranking criterion to keep track of skin barrier restoration after SM cutaneous intoxication in our SKH-1 mouse model. The R2 and R6 elasticity parameters or L° for the skin color exhibited their ability to be restored after 28 days of SM exposure. Our findings suggest that bio-engineering methods are eligible to evaluate new treatments on SM-induced skin SKH-1 mouse lesions, thus making an allowance for less invasive methods such as histological, genomic or proteomic approaches.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Anti-Matrix Metalloproteinase-9 antibody produced in goat, affinity isolated antibody