- Renal Angiotensinogen Is Predominantly Liver Derived in Nonhuman Primates.
Renal Angiotensinogen Is Predominantly Liver Derived in Nonhuman Primates.
Arteriosclerosis, thrombosis, and vascular biology (2021-09-10)
Masayoshi Kukida, Lei Cai, Dien Ye, Hisashi Sawada, Yuriko Katsumata, Michael K Franklin, Peter I Hecker, Kenneth S Campbell, A H Jan Danser, Adam E Mullick, Alan Daugherty, Ryan E Temel, Hong S Lu
PMID34496634
RÉSUMÉ
[Figure: see text].
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Anti-ACE antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-REN antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution