Merck
CTLA-4 expressed by FOXP3+ regulatory T cells prevents inflammatory tissue attack and not T-cell priming in arthritis.
Immunology (2017-05-13)
Katrin Klocke, Rikard Holmdahl, Kajsa Wing
RÉSUMÉ
Cytotoxic T-lymphocyte antigen 4 (CTLA-4) -mediated regulation of already tolerized autoreactive T cells is critical for understanding autoimmune responses. Although defects in CTLA-4 contribute to abnormal FOXP3+ regulatory T (Treg) cell function in rheumatoid arthritis, its role in autoreactive T cells remains elusive. We studied immunity towards the dominant collagen type II (CII) T-cell epitope in collagen-induced arthritis both in the heterologous setting and in the autologous setting where CII is mutated at position E266D in mouse cartilage. CTLA-4 regulated all stages of arthritis, including the chronic phase, and affected the priming of autologous but not heterologous CII-reactive T cells. CTLA-4 expression by both conventional T (Tconv) cells and Treg cells was required but while Tconv cell expression was needed to control the priming of naive autoreactive T cells, CTLA-4 on Treg cells prevented the inflammatory tissue attack. This identifies a cell-type-specific time window when CTLA-4-mediated tolerance is most powerful, which has important implications for clinical therapy with immune modulatory drugs.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Millipore
Plaque de 96 puits Multiscreen®, membrane en PVDF hydrophobe, pore size 0.45 μm, sterile