Merck
Swab-based enzyme immunoassay system for detection of meat residues on food contact surfaces as a hygiene monitoring tool.
Journal of food protection (1999-07-27)
B W Blais
RÉSUMÉ
An enzyme immunoassay system based on the use of a macroporous swab as a solid phase for the capture and subsequent immunoenzymatic detection of immunoglobulin G (IgG) from meat residues on food contact surfaces was developed as a hygiene-monitoring tool. Moistened polyester swabs coated with anti-bovine or anti-chicken IgG were rubbed on the test surface, and the captured IgG was subsequently detected directly on the swabs by brief sequential reactions with anti-bovine or anti-chicken IgG-peroxidase conjugate and chromogenic peroxidase substrate.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Anti-Bovine IgG (whole molecule) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution