Merck
BLT-1, a specific inhibitor of the HDL receptor SR-BI, induces a copper-dependent phenotype during zebrafish development.
Toxicology letters (2007-09-25)
Demetrio Raldúa, Patrick J Babin
RÉSUMÉ
Block lipid transport-1 (BLT-1) is a small chemical widely used to inhibit the transfer of lipids between high-density lipoproteins (HDL) and cells mediated by scavenger receptor B, type 1 (SR-BI). This study demonstrated that BLT-1 induced in zebrafish (Danio rerio) embryos a copper-dependent phenotype with a twisted notochord, brain ventricle enlargement, and absence of melanisation, phenocopying neocuproine-treated, or calamity mutants. This finding supports an unexpected link between copper availability and SR-BI activity. The copper-chelating activity of BLT-1, revealed by its dramatic effect during embryo development, should be considered in any evaluation of the pharmacological effect of this thiosemicarbazone derivative on SR-BI activity and the potential therapeutic value of this molecule.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
HDL Receptor SR-BI Inhibitor, BLT-1, The HDL Receptor SR-BI Inhibitor, BLT-1, also referenced under CAS 321673-30-7, controls the biological activity of HDL Receptor SR-B. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.