Merck
β-Cyclocitral is a conserved root growth regulator.
Proceedings of the National Academy of Sciences of the United States of America (2019-05-10)
Alexandra J Dickinson, Kevin Lehner, Jianing Mi, Kun-Peng Jia, Medhavinee Mijar, José Dinneny, Salim Al-Babili, Philip N Benfey
RÉSUMÉ
Natural compounds capable of increasing root depth and branching are desirable tools for enhancing stress tolerance in crops. We devised a sensitized screen to identify natural metabolites capable of regulating root traits in Arabidopsis β-Cyclocitral, an endogenous root compound, was found to promote cell divisions in root meristems and stimulate lateral root branching. β-Cyclocitral rescued meristematic cell divisions in ccd1ccd4 biosynthesis mutants, and β-cyclocitral-driven root growth was found to be independent of auxin, brassinosteroid, and reactive oxygen species signaling pathways. β-Cyclocitral had a conserved effect on root growth in tomato and rice and generated significantly more compact crown root systems in rice. Moreover, β-cyclocitral treatment enhanced plant vigor in rice plants exposed to salt-contaminated soil. These results indicate that β-cyclocitral is a broadly effective root growth promoter in both monocots and eudicots and could be a valuable tool to enhance crop vigor under environmental stress.