Merck
Identification of N-iminoethyl-L-ornithine as an irreversible inhibitor of nitric oxide synthase in phagocytic cells.
British journal of pharmacology (1991-01-01)
T B McCall, M Feelisch, R M Palmer, S Moncada
RÉSUMÉ
1. The synthesis of nitric oxide (NO) from L-arginine by rat peritoneal neutrophils (PMN) and the murine macrophage cell-line J774 and the inhibition of this synthesis by N-iminoethyl-L-ornithine (L-NIO), NG-monomethyl-L-arginine (L-NMMA), NG-nitro-L-arginine (L-NNA) and its methyl ester (L-NAME) were investigated. 2. L-NIO was the most potent inhibitor in both types of cells while L-NMMA was less active. L-NNA and L-NAME had no significant effect in PMN and L-NNA produced only approximately 40% inhibition of the generation of NO in the J774 cells at the highest concentration tested (300 microM). 3. The inhibitory effect of L-NIO was rapid in onset, requiring 10 min pre-incubation to achieve its full inhibitory activity, while the other compounds required 20-60 min pre-incubation to achieve their full effect. 4. The inhibitory effect of L-NIO (10 microM) on intact cells could not be reversed by L-arginine (300 microM) but could be prevented by concomitant incubation with this compound (300 microM), while the effect of the other inhibitors could be reversed by a 3-5 fold molar excess of L-arginine. 5. The NO synthase from both PMN and J774 cells was cytosolic and NADPH- but not Ca2(+)-dependent, with Km values for L-arginine of 3.3 +/- 0.8 and 4.2 +/- 1.1 microM respectively. 6. L-NIO was the most potent inhibitor of the neutrophil and J774 enzymes with IC50 values of 0.8 +/- 0.1 and 3 +/- 0.5 microM respectively. Furthermore, the effect of L-NIO was irreversible. The other three compounds were less potent, reversible inhibitors. 7. The inhibitory effects of all these compounds were enantiomerically specific. 8. These data indicate that L-NIO is a novel, potent, rapid in onset and irreversible inhibitor of NO synthase in phagocytic cells. The rapid uptake of L-NIO compared with the other compounds indicates that phagocytic cells have different uptake mechanisms for L-arginine analogues.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Nα-Methyl-L-ornithine monohydrochloride, ≥98% (TLC)