Merck
High-rate activated sludge systems combined with dissolved air flotation enable effective organics removal and recovery.
Bioresource technology (2019-07-30)
Cristina Cagnetta, Bart Saerens, Francis A Meerburg, Stijn O Decru, Eddie Broeders, Wilbert Menkveld, Tom G L Vandekerckhove, Jo De Vrieze, Siegfried E Vlaeminck, Arne R D Verliefde, Bart De Gusseme, Marjoleine Weemaes, Korneel Rabaey
RÉSUMÉ
High-rate activated sludge (HRAS) systems typically generate diluted sludge which requires further thickening prior to anaerobic digestion (AD), besides the need to add considerable coagulant and flocculant for the solids separation. As an alternative to conventional gravitational settling, a dissolved air flotation (DAF) unit was coupled to a HRAS system or a high-rate contact stabilization (HiCS) system. The HRAS-DAF system allowed up to 78% removal of the influent solids, and the HiCS-DAF 67%. Both were within the range of values typically obtained for HRAS-settler systems, albeit at a lower chemical requirement. The separated sludge had a high concentration of up to 47 g COD L-1, suppressing the need of further thickening before AD. Methanation tests showed a biogas yield of up to 68% on a COD basis. The use of a DAF separation system can thus enable direct organics removal at high sludge concentration and with low chemical needs.