Merck
Vasopressin regulates the growth of the biliary epithelium in polycystic liver disease.
Laboratory investigation; a journal of technical methods and pathology (2016-10-26)
Romina Mancinelli, Antonio Franchitto, Shannon Glaser, Antonella Vetuschi, Julie Venter, Roberta Sferra, Luigi Pannarale, Francesca Olivero, Guido Carpino, Gianfranco Alpini, Paolo Onori, Eugenio Gaudio
RÉSUMÉ
The neurohypophysial hormone arginine vasopressin (AVP) acts by three distinct receptor subtypes: V1a, V1b, and V2. In the liver, AVP is involved in ureogenesis, glycogenolysis, neoglucogenesis and regeneration. No data exist about the presence of AVP in the biliary epithelium. Cholangiocytes are the target cells in a number of animal models of cholestasis, including bile duct ligation (BDL), and in several human pathologies, such as polycystic liver disease characterized by the presence of cysts that bud from the biliary epithelium. In vivo, liver fragments from normal and BDL mice and rats as well as liver samples from normal and ADPKD patients were collected to evaluate: (i) intrahepatic bile duct mass by immunohistochemistry for cytokeratin-19; and (ii) expression of V1a, V1b and V2 by immunohistochemistry, immunofluorescence and real-time PCR. In vitro, small and large mouse cholangiocytes, H69 (non-malignant human cholangiocytes) and LCDE (human cholangiocytes from the cystic epithelium) were stimulated with vasopressin in the absence/presence of AVP antagonists such as OPC-31260 and Tolvaptan, before assessing cellular growth by MTT assay and cAMP levels. Cholangiocytes express V2 receptor that was upregulated following BDL and in ADPKD liver samples. Administration of AVP increased proliferation and cAMP levels of small cholangiocytes and LCDE cells. We found no effect in the proliferation of large mouse cholangiocytes and H69 cells. Increases were blocked by preincubation with the AVP antagonists. These results showed that AVP and its receptors may be important in the modulation of the proliferation rate of the biliary epithelium.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Anti-AVPR1A antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Le tarif et la disponibilité ne sont pas disponibles actuellement.