Z374938
Micro mats for PCR plates
About This Item
Produits recommandés
Technique(s)
PCR: suitable
Catégories apparentées
Description générale
- Fits most popular 96-well PCR plates
- Minimized evaporation in PCR and storage
- Alpha numerics printed on each mat, for easy well identification
- Pierceable with syringe
- Not compatible with DMSO and other harsh solvents
- Tabs for easy application/removal of mat
- Reusable for up to 5 times
- When used with a screw- or clip-down thermal cycler lid, provides 100% sealing
- Fully autoclavable, reversible
Les clients ont également consulté
Protocoles
REDAccuTaq LA protocol offers high-fidelity amplification of long PCR fragments with direct gel loading capability.
REDAccuTaq LA protocol offers high-fidelity amplification of long PCR fragments with direct gel loading capability.
REDAccuTaq LA protocol offers high-fidelity amplification of long PCR fragments with direct gel loading capability.
REDAccuTaq LA protocol offers high-fidelity amplification of long PCR fragments with direct gel loading capability.