Key Documents
CLS9186
Corning® aspiration pipettes
volume 2 mL
Synonyme(s) :
Aspiration pipette, Corning pipette
About This Item
Code UNSPSC :
41121500
eCl@ss :
32070201
Nomenclature NACRES :
NB.01
Matériaux
polystyrene
Stérilité
sterile
Conditionnement
pack of 100 ea
case of 1000 ea
Fabricant/nom de marque
Corning 9186
Capacité
2 mL
Volume
2 mL
Description générale
  • Corning® aspirating pipettes are made of optically clear polystyrene for durability and good mechanical strength.
  • The sterile, ungraduated, and unplugged pipettes are best suited for aspirating liquid aseptically using vacuum suction.
  • Individually wrapped and bulk packed.
  • Disposable and certified non-pyrogenic.
Aspirating pipettes are sterile, ungraduated and unplugged polystyrene pipettes for aspirating liquid using vacuum suction.
Informations légales
Corning is a registered trademark of Corning, Inc.