Key Documents
BR43238
BRAND® density bottle, Gay-Lussac pattern, uncalibrated
capacity 100 mL, borosilicate glass
Synonyme(s) :
Gay-Lussac
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.71
Matériaux
borosilicate glass
glass stopper
narrow-mouth bottle
round bottle (Gay-Lussac)
white bottle
Joint
joint ST/NS neck10/19
Caractéristiques
with cap
Conditionnement
pack of 2 ea
Fabricant/nom de marque
BRAND 43238
Capacité
100 mL
Description générale
DIN ISO 3507, type Gay-Lussac. Stopper NS 10/19 with capillary. Top of stopper ground and polished. Nominal capacity printed on the bottom.
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG
Protocoles
Density bottles determine liquid density, calibrated 'to contain' for accuracy, though not volumetric instruments like volumetric flasks.
Density bottles determine liquid density, calibrated 'to contain' for accuracy, though not volumetric instruments like volumetric flasks.
Density bottles determine liquid density, calibrated 'to contain' for accuracy, though not volumetric instruments like volumetric flasks.
Density bottles determine liquid density, calibrated 'to contain' for accuracy, though not volumetric instruments like volumetric flasks.