Key Documents
BMSC2570
Benchmark Centrifuge Adapter 15 mL rotor to 5 mL tubes
Synonyme(s) :
tube adapter
About This Item
Code UNSPSC :
41103911
Nomenclature NACRES :
NB.82
Fabricant/nom de marque
Benchmark Scientific C2570