Merck
Novel role of surfactant protein A in bacterial sinusitis.
International forum of allergy & rhinology (2017-07-21)
George T Noutsios, Amanda L Willis, Julie G Ledford, Eugene H Chang
ABSTRACT
Chronic rhinosinusitis (CRS) is a common inflammatory disorder of the upper airway characterized by chronic inflammation and significant sinonasal remodeling. CRS is comprised of 2 major subgroups, based on whether polyps are present or absent. In some cases, it is characterized by colonization with opportunistic pathogens such as Pseudomonas aeruginosa (PA), Staphylococcus aureus, and other bacteria. The innate immune system of the sinonasal epithelium is the first line of defense against inhaled pathogens. Surfactant protein A (SP-A) is a member of the collectin family secreted by the airway epithelia and plays a critical role in airway innate immunity, as it can aggregate bacteria. We hypothesized that SP-A plays a role in bacterial CRS. Air-liquid interface (ALI) cultures of nasal epithelial cells were derived from human ex-vivo healthy and CRS sinus tissues (n = 26) and challenged with PA. SP-A levels were measured with western blot and quantitative reverse transcript-polymerase chain reaction (qRT-PCR) in ALI and sinus tissues. We determined that SP-A: (i) mRNA and protein levels are increased significantly in CRS tissues compared with healthy sinuses; (ii) although primarily expressed in the lung, it is also synthesized and expressed in sinonasal epithelia; (ii) is expressed in the sinuses of an SP-A humanized transgenic mouse but not in SP-A knockout mice; (iv) mRNA levels are upregulated significantly during PA challenge, but protein levels are downregulated 4 hours postchallenge and upregulated at 12 hours. Our data suggest that SP-A is expressed in the sinuses and that it plays a role in the sinus innate immune responses during bacterial infections.
Sigma-Aldrich
Anti-Surfactant Protein A Antibody, from rabbit
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O