Merck
  • Midgut serine proteases and alternative host plant utilization in Pieris brassicae L.
Midgut serine proteases and alternative host plant utilization in Pieris brassicae L.
Frontiers in physiology (2015-04-16)
Rakesh Kumar, Usha Bhardwaj, Pawan Kumar, Sudeshna Mazumdar-Leighton
ABSTRACT
Pieris brassicae L. is a serious pest of cultivated crucifers in several parts of the world. Larvae of P. brassicae also feed prolifically on garden nasturtium (Tropaeolum majus L., of the family Tropaeolaceae). Proteolytic digestion was studied in larvae feeding on multiple hosts. Fourth instars were collected from cauliflower fields before transfer onto detached, aerial tissues of selected host plants in the lab. Variable levels of midgut proteases were detected in larvae fed on different hosts using protein substrates (casein and recombinant RBCL cloned from cauliflower) and diagnostic, synthetic substrates. Qualitative changes in midgut trypsin activities and quantitative changes in midgut chymotrypsin activities were implicated in physiological adaptation of larvae transferred to T. majus. Midgut proteolytic activities were inhibited to different extents by serine protease inhibitors, including putative trypsin inhibitors isolated from herbivore-attacked and herbivore-free leaves of cauliflower (CfTI) and T. majus (TpTI). Transfer of larvae to T. majus significantly influenced feeding parameters but not necessarily when transferred to different tissues of the same host. Results obtained are relevant for devising sustainable pest management strategies, including transgenic approaches using genes encoding plant protease inhibitors.
Sigma-Aldrich
Nα-p-Tosyl-L-arginine methyl ester hydrochloride
Sigma-Aldrich
Nα-Benzoyl-L-arginine-7-amido-4-methylcoumarin hydrochloride, protease substrate
Roche
Aprotinin, from bovine lung