Merck
  • cAMP signaling increases histone deacetylase 8 expression by inhibiting JNK-dependent degradation via autophagy and the proteasome system in H1299 lung cancer cells.
cAMP signaling increases histone deacetylase 8 expression by inhibiting JNK-dependent degradation via autophagy and the proteasome system in H1299 lung cancer cells.
Biochemical and biophysical research communications (2016-01-23)
Ji-Yeon Park, Yong-Sung Juhnn
ABSTRACT
This study aimed to investigate the roles of autophagy and the ubiquitin-proteasome system in the degradation of histone deacetylase 8 (HDAC8) and to clarify the mechanism by which cAMP signaling regulates this degradation. cAMP signaling was activated by treating H1299 non-small cell lung cancer cells with isoproterenol or forskolin/3-isobutyl-1-methylxanthine, and HDAC8 expression was assessed by western blot analysis. The inhibition of autophagy and ubiquitin-proteasome-dependent degradation increased HDAC8 expression. cAMP signaling inhibited JNK activation, which decreased the phosphorylation of Bcl-2, thereby reducing autophagy, and the phosphorylation of Itch, thereby reducing ubiquitination. These results suggest that the HDAC8 protein is degraded via autophagy and the ubiquitin-proteasome system and that cAMP signaling increases HDAC8 protein levels by reducing JNK-mediated autophagy and ubiquitin-proteasome-dependent degradation of the HDAC8 protein in H1299 lung cancer cells.
Sigma-Aldrich
PYR-41, ≥98% (HPLC), powder
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
SP600125, ≥98% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
MISSION® esiRNA, targeting human ITCH