Merck
  • Five new diarylheptanoids from the rhizomes of Curcuma kwangsiensis and their antiproliferative activity.
Five new diarylheptanoids from the rhizomes of Curcuma kwangsiensis and their antiproliferative activity.
Fitoterapia (2015-02-24)
Shao-Dan Chen, Jin-Tao Gao, Jing-Gong Liu, Bo Liu, Rui-Zhi Zhao, Chuan-Jian Lu
ABSTRACT
Five new diarylheptanoids (1-5), along with nine known ones (6-14), were isolated from the rhizomes of Curcuma kwangsiensis. Their structures were established on the basis of spectroscopic analyses. Compounds 1-3 were cyclic diarylheptanoids rarely discovered from C. kwangsiensis. Of all the isolated compounds, compound 4 showed moderate antiproliferative activity on HH and HaCaT cells.
Sigma-Aldrich
Pyridine-d5, ≥99.5 atom % D
Sigma-Aldrich
Pyridine-d5, "100%", ≥99.96 atom % D
Sigma-Aldrich
Pyridine-d5, ≥99.5 atom % D, contains 0.03 % (v/v) TMS