Merck
  • Interleukin 7 plays a role in T lymphocyte apoptosis inhibition driven by mesenchymal stem cell without favoring proliferation and cytokines secretion.
Interleukin 7 plays a role in T lymphocyte apoptosis inhibition driven by mesenchymal stem cell without favoring proliferation and cytokines secretion.
PloS one (2014-09-04)
Marilia Normanton, Heliene Alvarenga, Nelson Hamerschlak, Andreza Ribeiro, Andrea Kondo, Luiz Vicente Rizzo, Luciana Cavalheiro Marti
ABSTRACT
Since 2004, when a case report describing the use of human mesenchymal stem cells (hMSCs) infusion as a therapy for GVHD after bone marrow transplantation, a new perspective in MSC function emerged. Since then hMSCs immunomodulatory potential became the target of several studies. Although great progress has been made in our understanding of hMSCs, their effect on T cell remains obscure. Our study has confirmed the already described effect of hMSCs on lymphocytes proliferation and survival. We also show that the impairment of lymphocyte proliferation and apoptosis is contact-independent and occurs in a prostaglandin-independent manner. A potential correlation between IL-7 and hMSCs effect is suggested, as we observed an increase in IL-7 receptors (CD127) on lymphocyte membrane in MSC presence. Additionally, blocking IL-7 in hMSCs-lymphocytes co-cultures increased lymphocytes apoptosis and we also have demonstrated that hMSCs are able to produce this interleukin. Moreover, we found that during Th1/Th17 differentiation in vitro, hMSCs presence leads to Th1/Th17 cells with reduced capacity of INF-y and IL-17 secretion respectively, regardless of having several pro-inflammatory cytokines in culture. We did not confirm an increment of Treg in these cultures, but a reduced percentage of INF-y/IL-17 secreting cells was observed, suggesting that the ratio between anti and pro-inflammatory cells changed. This changed ratio is very important to GvHD therapy and links hMSCs to an anti-inflammatory role. Taken together, our findings provide important preliminary results on the lymphocyte pathway modulated by MSCs and may contribute for developing novel treatments and therapeutic targets for GvHD and others autoimmune diseases.
Sigma-Aldrich
IL-4 human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)
Sigma-Aldrich
IL-4 human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
Interleukin-4 from mouse, IL-4, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Dinoprostone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Interleukin-4 human, IL-4, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Anti-IL7 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Interleukin-4 human, IL-4, potency:, recombinant, expressed in HEK 293 cells, HumanKine®, suitable for cell culture
Sigma-Aldrich
IL-4 from mouse, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Interleukin-4 from rat, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture, ≥97% (SDS-PAGE)
Sigma-Aldrich
Prostaglandin E2, synthetic, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Prostaglandin E2, ≥93% (HPLC), synthetic
Sigma-Aldrich
Prostaglandin E2, γ-irradiated, powder, BioXtra, suitable for cell culture