Merck
Bupropion hydrochloride.
Drug intelligence & clinical pharmacy (1984-12-01)
R L Dufresne, S S Weber, R E Becker
ABSTRACT
Bupropion is a trimethylated monocyclic phenylaminoketone that is an effective antidepressant in humans. It neither is sedating, anticholinergic, nor cardiotoxic. Its mechanism of action may be related to dopamine, but remains uncertain at this time. Clinical trials comparing bupropion 300-750 mg/d with placebo show it to be superior to placebo in efficacy and as well tolerated. Bupropion, in controlled clinical trials, is as effective as amitriptyline or imipramine, with fewer side effects. The only clinically significant adverse reaction to bupropion in more than 1000 patients studied has been seizure induction at a frequency comparable with that of imipramine. Bupropion appears to be safe and effective in both adult and geriatric depressed patients. Although it appears to be safer and equally efficacious when compared with currently used antidepressants, it has not been tested by routine clinical use.
Product Description
Supelco
Bupropion hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
USP
Bupropion hydrochloride, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Bupropion hydrochloride, ≥98% (HPLC), solid