Merck
Lamotrigine in psychiatric disorders.
The Journal of clinical psychiatry (2013-08-16)
Jennifer G Reid, Michael J Gitlin, Lori L Altshuler
ABSTRACT
Owing to the prevalence of medication side effects and treatment resistance, prescribers often consider off-label uses of US Food and Drug Administration (FDA)-approved agents for the treatment of persistent symptoms. The authors review the available literature on the FDA-approved and non-FDA-approved uses of lamotrigine in adults with psychiatric disorders. We used PubMed, MEDLINE, and a hand search of relevant literature to find studies published between 1990 and 2012 and available in English language. The following keywords were searched: lamotrigine, psychiatric, mood disorders, depression, personality disorders, anxiety, schizophrenia, side effects, and rash. Data were selected from 29 randomized controlled trials (RCTs). When RCTs were not available, open-label trials (6), retrospective case reviews (10), and case series (4) were summarized. We extracted results of monotherapy and augmentation trials of lamotrigine on primary and secondary outcome measures. Lamotrigine is generally well tolerated, with the best evidence for the maintenance treatment of bipolar disorder, particularly in prevention of depressive episodes. In acute bipolar depression, meta-analyses suggested a modest benefit, especially for more severely depressed subjects, with switch rates similar to placebo. In unipolar depression, double-blind RCTs noted benefit on subsets of symptoms and improved response in more severely depressed subjects. Data are limited but promising in borderline personality disorder. Use of lamotrigine in schizophrenia and anxiety disorders has little supportive evidence. Lamotrigine is recommended in bipolar maintenance when depression is prominent. It also has a role in treating acute bipolar depression and unipolar depression, though the latter warrants more research. Data are too limited in other psychiatric disorders to recommend its use at this time.
Product Description
Supelco
Lamotrigine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Lamotrigine, ≥98%, powder
Lamotrigine for peak identification, European Pharmacopoeia (EP) Reference Standard
Lamotrigine, European Pharmacopoeia (EP) Reference Standard
Lamotrigine for system suitability, European Pharmacopoeia (EP) Reference Standard
USP
Lamotrigine, United States Pharmacopeia (USP) Reference Standard
Supelco
Lamotrigine, Pharmaceutical Secondary Standard; Certified Reference Material