Merck
Urine nandrolone metabolites: false positive doping test?
British journal of sports medicine (2002-09-28)
R M N Kohler, M I Lambert
ABSTRACT
The aim of this review is to analyse the studies on nandrolone metabolism to determine if it is possible for an athlete to test positive for nandrolone without having ingested or injected nandrolone.
Sigma-Aldrich
19-Nortestosterone
Sigma-Aldrich
19-Nortestosterone, ≥99.0% (HPLC)
Supelco
Nandrolone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®