Merck
  • [Studies of chemopreventive agents against neoplasma: synthesis of 3-acetyl coumarin derivatives and relationship between antimutagenic activity and structure].
[Studies of chemopreventive agents against neoplasma: synthesis of 3-acetyl coumarin derivatives and relationship between antimutagenic activity and structure].
Yao xue xue bao = Acta pharmaceutica Sinica (1996-01-01)
X L Huang, S P Xu, Z D Fu, B An
ABSTRACT
Twenty-five 3-acetylcoumarin derivatives were synthesized among which twenty-two were not reported before. Antimutagenic activity screen in vitro has shown that some of these compounds have various activities. The structure and activity relationship for 5-, 7-, 8-substituents has been studied. Pharmacological data showed that: the substituent on position 8 has important effect on its activity. When there is only a hydroxy group on position 7, its activity is the highest among those with other substituents, but when a methyl is on position 8, the order of the activity is reversed. Other trends have also been found which provided some clues for further structural modification.
Sigma-Aldrich
3-Acetylcoumarin, 96%