Merck
  • Cytotoxic aspects of gadolinium oxide nanostructures for up-conversion and NIR bioimaging.
Cytotoxic aspects of gadolinium oxide nanostructures for up-conversion and NIR bioimaging.
Acta biomaterialia (2012-09-12)
Eva Hemmer, Tomoyoshi Yamano, Hidehiro Kishimoto, Nallusamy Venkatachalam, Hiroshi Hyodo, Kohei Soga
ABSTRACT
Bioimaging is an important diagnostic tool in the investigation and visualization of biological phenomena in cells and in medicine. In this context, up-converting Gd(2)O(3):Er(3+),Yb(3+) nanostructures (nanoparticles, nanorods) have been synthesized by precipitation methods and hydrothermal synthesis. Independent of size and morphology, Gd(2)O(3):Er(3+),Yb(3+) powders show up-conversion (550 nm, 670 nm) and near-infrared emission (1.5 μm) upon 980 nm excitation, which makes these structures interesting for application as biomarkers. With regard to their potential application in bioimaging, cytotoxicity is an important aspect and is strongly affected by the physico-chemical properties of the investigated nanostructures. Therefore, the cytotoxic effect of bare and poly(ethylene glycol)-b-poly(acrylic acid) block co-polymer-modified nanostructures on non-phagocytic and phagocytic cells (B-cell hybridoma cells and macrophages) was investigated. The observed cytotoxic behavior in the case of macrophages incubated with bare nanostructures was assigned to the poor chemical durability of gadolinium oxide, but could be overcome by surface modification.
Sigma-Aldrich
Gadolinium(III) oxide, powder, 99.9% trace metals basis
Sigma-Aldrich
Gadolinium(III) oxide, ≥99.9%
Sigma-Aldrich
Gadolinium(III) oxide, powder, ≥99.99% trace metals basis
Sigma-Aldrich
Gadolinium(III) oxide, nanopowder, <100 nm particle size (BET), 99.8% trace metals basis