Merck
  • Topotecan exposure estimation in pediatric acute myeloid leukemia supported by LC-MS-based drug monitoring and pharmacokinetic analysis.
Topotecan exposure estimation in pediatric acute myeloid leukemia supported by LC-MS-based drug monitoring and pharmacokinetic analysis.
Journal of pharmaceutical and biomedical analysis (2012-06-05)
Tomasz Bączek, Lucyna Konieczna, Mariusz Belka, Lucyna Maciejka-Kapuścińska, Jakub Wiśniewski, Maciej Niedźwiecki, Anna Balcerska, Elżbieta Adamkiewicz-Drożyńska, Jacek Wachowiak
ABSTRACT
Individualization of the topotecan dosing can reduce inter-patient variability, toxicity, and at the same time increases chemotherapy efficacy. Topotecan dosing based on simultaneous drug monitoring and pharmacokinetic analysis can yield more accurate and precise estimation of the topotecan systemic exposure than that attainable with the fixed dosing approach. Therefore, a combined approach could provide a tool assisting the clinicians in individualization of the topotecan dosing. The aim of the study was to estimate the topotecan exposure in pediatric patients with acute myeloid leukemia (AML) based on the plasma concentration-time data and using the pharmacokinetic analysis. The primary goal was achieve the correct estimation of the target plasma area against the topotecan concentration-time curve (AUC) in a 5 day course of cladribine followed by monitored topotecan in pediatric patients with recurrent/refractory AML. A sensitive and selective reversed-phase liquid chromatographic-mass spectrometry (LC-MS) assay was developed to quantify total topotecan in the human plasma samples. This method, with its lower quantification limit of 1 ng/ml, was validated over a linear range of 1-150 ng/ml. Under the proposed approach, the topotecan dosing was selected so as to achieve the final AUC value of 140±20 ng/ml h. The presented analytical and pharmacokinetic data demonstrate that the proposed approach can be a practical, useful, efficient, and accurate tool for individualizing the topotecan dosing in children with AML.
Sigma-Aldrich
2-Chloro-2′-deoxyadenosine, antileukemic
Pricing and availability is not currently available.
Cladribine for peak identification, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.