Merck
  • [Study on rat nasal absorption in situ of borneol based on single pass perfusion method].
[Study on rat nasal absorption in situ of borneol based on single pass perfusion method].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2012-01-20)
Xiao Song, Shouying Du, Yang Lu, Yong Ma, Xiaolan Chen, Yue Wang, Huixian Zhang
ABSTRACT
To investigate the absorption characteristic of borneol. Using single pass perfusion model, the active ingredient of borneol were detected by GC. The drug concentration, perfusion rate and pH value on the absorption of borneol were studied. Perfusion rate on the absorption rate constants (Ka) had significant effects. Drug concentration and pH value on the absorption rate constants had no significant impact. the absorption of borneol is good by nasal. The absorption rate constants of borneol have no effected by drug concentration. The absorption of borneol is via a simple diffusion.
Product Description
Supelco
(+)-Borneol, analytical standard
Sigma-Aldrich
(+)-Borneol, 97%
Supelco
(−)-Borneol, analytical standard
Sigma-Aldrich
Isoborneol, 95%
Sigma-Aldrich
(−)-Borneol, predominantly endo, 97%