Merck
  • Molecular and biochemical characterization of the tetralin degradation pathway in Rhodococcus sp. strain TFB.
Molecular and biochemical characterization of the tetralin degradation pathway in Rhodococcus sp. strain TFB.
Microbial biotechnology (2009-03-01)
Laura Tomás-Gallardo, Eduardo Santero, Emilio Camafeita, Enrique Calvo, Michael Schlömann, Belén Floriano
ABSTRACT
The tetralin biodegradation pathway in Rhodococcus sp. strain TFB, a Gram-positive bacterium resistant to genetic manipulation, was characterized using a proteomic approach. Relative protein expression in cell free extracts from tetralin- and glucose-grown cells was compared using the 2D-DIGE technique. Identification of proteins specifically expressed in tetralin-grown cells was used to characterize a complete set of genes involved in tetralin degradation by reverse genetics. We propose a tetralin degradation pathway analogous to that described for Sphingomonas macrogolitabida strain TFA. TFB thn genes are organized into three operons; two contain all of the structural genes and are transcribed in the same direction, while the third operon, thnST, is transcribed in the opposite direction and encodes a two-component regulatory system, whose transcription is higher in tetralin-grown cells. In addition to tetralin induction, TFB thn structural genes are subject to glucose repression. Primer extension assays and translational thnA1::gfp and thnS::gfp fusions were used to characterize putative promoter regions. A mutational analysis of the thnA1 promoter region allowed us to define nucleotides within the cis regulatory elements that are important for the control of thn gene expression.
Sigma-Aldrich
1,2,3,4-Tetrahydronaphthalene, anhydrous, 99%
Supelco
1,2,3,4-Tetrahydronaphthalene, analytical standard
Sigma-Aldrich
1,2,3,4-Tetrahydronaphthalene, ReagentPlus®, 99%
Sigma-Aldrich
1,2,3,4-Tetrahydronaphthalene, reagent grade, ≥97%