Merck
Urinary metabolites of di-n-octyl phthalate in rats.
Toxicology (2005-04-21)
Manori J Silva, Kayoko Kato, Earl L Gray, Cynthia Wolf, Larry L Needham, Antonia M Calafat
ABSTRACT
Di-n-octyl phthalate (DnOP) is a plasticizer used in polyvinyl chloride plastics, cellulose esters, and polystyrene resins. The metabolism of DnOP results in the hydrolysis of one ester linkage to produce mono-n-octyl phthalate (MnOP), which subsequently metabolizes to form oxidative metabolites. We investigated the toxicokinetics of DnOP in adult female Sprague-Dawley rats by monitoring the excretion of DnOP metabolites in urine after oral administration of DnOP (300 mg/kg). By using authentic standards, the presence of urinary phthalic acid (PA), MnOP, and the major DnOP metabolite, mono-(3-carboxypropyl) phthalate (MCPP) was clearly established. Furthermore, we identified five additional urinary DnOP oxidative metabolites based on their chromatographic behavior and mass spectrometric fragmentation pattern. These DnOP oxidative metabolites, are postulated to be mono-carboxymethyl phthalate (MCMP), mono-(5-carboxy-n-pentyl) phthalate (MCPeP), mono-(7-carboxy-n-heptyl) phthalate (MCHpP), and isomers of mono-hydroxy-n-octyl phthalate (MHOP) (e.g., mono-(7-hydroxy-n-octyl) phthalate) and of mono-oxo-n-octyl phthalate (MOOP) (e.g., mono-(7-oxo-n-octyl) phthalate). The urinary excretion of DnOP metabolites followed a biphasic excretion pattern. The metabolite levels decreased significantly after the first day of DnOP administration although MCPP, MCHpP, MHOP, and MOOP were detectable after 4 days. We also studied the in vitro metabolism of DnOP and MnOP by rat liver microsomes. DnOP produced MnOP, MHOP, and PA in vitro whereas, MnOP produced MHOP and PA in vitro at detectable levels.
Sigma-Aldrich
Di-n-octyl phthalate, ≥98.0% (GC)
Pricing and availability is not currently available.
Supelco
Di-n-octyl phthalate, analytical standard
Pricing and availability is not currently available.
Supelco
Di-n-octyl phthalate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Pricing and availability is not currently available.