Merck
Integral and associated lysosomal membrane proteins.
Traffic (Copenhagen, Denmark) (2007-09-28)
Bernd Schröder, Christian Wrocklage, Cuiping Pan, Ralf Jäger, Bernd Kösters, Helmut Schäfer, Hans-Peter Elsässer, Matthias Mann, Andrej Hasilik
ABSTRACT
We searched for novel proteins in lysosomal membranes, tentatively participating in molecular transport across the membrane and/or in interactions with other compartments. In membranes purified from placental lysosomes, we identified 58 proteins, known to reside at least partially in the lysosomal membrane. These included 17 polypeptides comprising or associated with the vacuolar adenosine triphosphatase. We report on additional 86 proteins that were significantly enriched in the lysosomal membrane fraction. Among these, 12 novel proteins of unknown functions were found. Three were orthologues of rat proteins that have been identified in tritosomes by Bagshaw RD et al. (A proteomic analysis of lysosomal integral membrane proteins reveals the diverse composition of the organelle. Mol Cell Proteomics 2005;4:133-143). Here, the proteins encoded by LOC201931 (FLJ38482) and LOC51622 (C7orf28A) were expressed with an appended fluorescent tag in HeLa cells and found to be present in lysosomal organelles. Among the lysosomally enriched proteins, also 16 enzymes and transporters were detected that had not been assigned to lysosomal membranes previously. Finally, our results identified a particular set of proteins with known functions in signaling and targeting to be at least partially associated with lysosomes.
Sigma-Aldrich
Phosphatase, Acid from sweet potato, ammonium sulfate suspension, ≥10.0 units/mg protein (modified Warburg-Christian)
Sigma-Aldrich
Phosphatase, Acid from potato, lyophilized powder, ≥0.5 unit/mg solid
Sigma-Aldrich
Phosphatase, Acid from potato, lyophilized powder, ≥3.0 units/mg solid
Sigma-Aldrich
5′-Nucleotidase human, recombinant, expressed in CHO cells, vial of 6-12 μg
Sigma-Aldrich
Phosphatase, Acid from wheat germ, ≥0.4 unit/mg solid