Merck
  • Extension of nasal anti-Staphylococcus aureus efficacy of lysostaphin by its incorporation into a chitosan-o/w cream.
Extension of nasal anti-Staphylococcus aureus efficacy of lysostaphin by its incorporation into a chitosan-o/w cream.
Drug delivery (2010-08-24)
Fuying Cui, Guodong Li, Jinjiang Huang, Jien Zhang, Min Lu, Wanying Lu, Qingshan Huang
ABSTRACT
Nasal colonization of Staphylococcus aureus (S.aureus) is known as a significant risk factor for nosocomial infections, and clearance of its nasal colonization greatly reduces the risk. In the present study the preparation and characterizations of the chitosan-o/w cream incorporated with lysostaphin (CCL) were described. It showed that the concentration of incorporated lysostaphin had a direct relationship with its release behavior from the cream. It was rapid at 2 and 3.5 mg lysostaphin/g cream and of a more sustained pattern at 5 mg lysostaphin/g cream. Efficacy of lysostaphin released from the CCL cream to inhibit S.aureus growth was higher than that of lysostaphin delivery routinely treated, as demonstrated by the reduction of the mucociliary transport rate (MTR) in contrast to the control graphite particles (p < 0.05). Therefore, it is concluded that drug delivery by the CCL holds its potential as a local nasal anti-S.aureus infection.
Sigma-Aldrich
Lysostaphin from Staphylococcus simulans, recombinant, expressed in E. coli, lyophilized powder
Sigma-Aldrich
Lysostaphin from Staphylococcus staphylolyticus, lyophilized powder, Protein 50-70 % by biuret, ≥500 units/mg protein
Sigma-Aldrich
Lysostaphin from Staphylococcus staphylolyticus, BioUltra, ≥97% (SDS-PAGE), Protein 40-60 % by biuret, ≥2,000 units/mg protein
Sigma-Aldrich
Lysostaphin from Staphylococcus staphylolyticus, aseptically filled