Merck
  • p57Kip2 acts as a transcriptional corepressor to regulate intestinal stem cell fate and proliferation.
p57Kip2 acts as a transcriptional corepressor to regulate intestinal stem cell fate and proliferation.
Cell reports (2023-06-16)
Justine Creff, Ada Nowosad, Anne Prel, Anne Pizzoccaro, Marion Aguirrebengoa, Nicolas Duquesnes, Caroline Callot, Thomas Jungas, Christine Dozier, Arnaud Besson
ABSTRACT
p57Kip2 is a cyclin/CDK inhibitor and a negative regulator of cell proliferation. Here, we report that p57 regulates intestinal stem cell (ISC) fate and proliferation in a CDK-independent manner during intestinal development. In the absence of p57, intestinal crypts exhibit an increased proliferation and an amplification of transit-amplifying cells and of Hopx+ ISCs, which are no longer quiescent, while Lgr5+ ISCs are unaffected. RNA sequencing (RNA-seq) analyses of Hopx+ ISCs show major gene expression changes in the absence of p57. We found that p57 binds to and inhibits the activity of Ascl2, a transcription factor critical for ISC specification and maintenance, by participating in the recruitment of a corepressor complex to Ascl2 target gene promoters. Thus, our data suggest that, during intestinal development, p57 plays a key role in maintaining Hopx+ ISC quiescence and repressing the ISC phenotype outside of the crypt bottom by inhibiting the transcription factor Ascl2 in a CDK-independent manner.
Sigma-Aldrich
Anti-p57Kip2 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-HDAC7 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-Achaete Scute homolog 2 Antibody, clone 8F1, clone 8F1, from mouse
Sigma-Aldrich
ANTI-ASCL2 (N-TERM) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution