Merck
  • Fibrocytes boost tumor-supportive phenotypic switches in the lung cancer niche via the endothelin system.
Fibrocytes boost tumor-supportive phenotypic switches in the lung cancer niche via the endothelin system.
Nature communications (2022-10-15)
Andreas Weigert, Xiang Zheng, Alina Nenzel, Kati Turkowski, Stefan Günther, Elisabeth Strack, Evelyn Sirait-Fischer, Eiman Elwakeel, Ivan M Kur, Vandana S Nikam, Chanil Valasarajan, Hauke Winter, Alexander Wissgott, Robert Voswinkel, Friedrich Grimminger, Bernhard Brüne, Werner Seeger, Soni Savai Pullamsetti, Rajkumar Savai
ABSTRACT
Fibrocytes are bone marrow-derived monocytic cells implicated in wound healing. Here, we identify their role in lung cancer progression/ metastasis. Selective manipulation of fibrocytes in mouse lung tumor models documents the central role of fibrocytes in boosting niche features and enhancing metastasis. Importantly, lung cancer patients show increased number of circulating fibrocytes and marked fibrocyte accumulation in the cancer niche. Using double and triple co-culture systems with human lung cancer cells, fibrocytes, macrophages and endothelial cells, we substantiate the central features of cancer-supporting niche: enhanced cancer cell proliferation and migration, macrophage activation, augmented endothelial cell sprouting and fibrocyte maturation. Upregulation of endothelin and its receptors are noted, and dual endothelin receptor blockade suppresses all cancer-supportive phenotypic alterations via acting on fibrocyte interaction with the cancer niche. We thus provide evidence for a crucial role of fibrocytes in lung cancer progression and metastasis, suggesting targets for treatment strategies.
Sigma-Aldrich
Anti-CD14 Antibody, clone 2D-15C, clone 2D-15C, Chemicon®, from mouse
Sigma-Aldrich
Anti-CD44 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Actin, α-Smooth Muscle - FITC antibody, Mouse monoclonal, clone 1A4, purified from hybridoma cell culture
Sigma-Aldrich
Collagen Type I, rat tail
Sigma-Aldrich
Anti-Human B cells (CD19) Antibody, clone FMC63, clone FMC63, Chemicon®, from mouse