Merck
  • Epstein-Barr virus BNRF1 destabilizes SMC5/6 cohesin complexes to evade its restriction of replication compartments.
Epstein-Barr virus BNRF1 destabilizes SMC5/6 cohesin complexes to evade its restriction of replication compartments.
Cell reports (2022-03-10)
Stephanie Pei Tung Yiu, Rui Guo, Cassie Zerbe, Michael P Weekes, Benjamin E Gewurz
ABSTRACT
Epstein-Barr virus (EBV) persistently infects people worldwide. Delivery of ∼170-kb EBV genomes to nuclei and use of nuclear membrane-less replication compartments (RCs) for their lytic cycle amplification necessitate evasion of intrinsic antiviral responses. Proteomics analysis indicates that, upon B cell infection or lytic reactivation, EBV depletes the cohesin SMC5/6, which has major roles in chromosome maintenance and DNA damage repair. The major tegument protein BNRF1 targets SMC5/6 complexes by a ubiquitin proteasome pathway dependent on calpain proteolysis and Cullin-7. In the absence of BNRF1, SMC5/6 associates with R-loop structures, including at the viral lytic origin of replication, and interferes with RC formation and encapsidation. CRISPR analysis identifies RC restriction roles of SMC5/6 components involved in DNA entrapment and SUMOylation. Our study highlights SMC5/6 as an intrinsic immune sensor and restriction factor for a human herpesvirus RC and has implications for the pathogenesis of EBV-associated cancers.
Sigma-Aldrich
Calpeptin, ≥98% (HPLC)
Sigma-Aldrich
Anti-DNA-RNA Hybrid Antibody, clone S9.6, clone S9.6, from mouse
Sigma-Aldrich
Anti-dsRNA Antibody, clone rJ2, culture supernatant, clone rJ2, from mouse
Roche
DNase I, grade II, from bovine pancreas
Sigma-Aldrich
E-64 Protease Inhibitor, The E-64 Protease Inhibitor, also referenced under CAS 66701-25-5, controls the biological activity of E-64 Protease. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.