Merck
  • Synergistic effect of non-neutralizing antibodies and interferon-γ for cross-protection against influenza.
Synergistic effect of non-neutralizing antibodies and interferon-γ for cross-protection against influenza.
iScience (2021-10-09)
Meito Shibuya, Shigeyuki Tamiya, Atsushi Kawai, Toshiro Hirai, Mark S Cragg, Yasuo Yoshioka
ABSTRACT
Current influenza vaccines do not typically confer cross-protection against antigenically mismatched strains. To develop vaccines conferring broader cross-protection, recent evidence indicates the crucial role of both cross-reactive antibodies and viral-specific CD4+ T cells; however, the precise mechanism of cross-protection is unclear. Furthermore, adjuvants that can efficiently induce cross-protective CD4+ T cells have not been identified. Here we show that CpG oligodeoxynucleotides combined with aluminum salts work as adjuvants for influenza vaccine and confer strong cross-protection in mice. Both cross-reactive antibodies and viral-specific CD4+ T cells contributed to cross-protection synergistically, with each individually ineffective. Furthermore, we found that downregulated expression of Fcγ receptor IIb on alveolar macrophages due to IFN-γ secreted by viral-specific CD4+ T cells improves the activity of cross-reactive antibodies. Our findings inform the development of optimal adjuvants for vaccines and how influenza vaccines confer broader cross-protection.
Sigma-Aldrich
Goat Anti-Mouse IgG γ chain Antibody, HRP conjugate, Species Adsorbed, Chemicon®, from goat
Sigma-Aldrich
Brefeldin A, ≥99% (HPLC and TLC), BioXtra, for molecular biology