Merck
  • A Seasonal Study of Koi Herpesvirus and Koi Sleepy Disease Outbreaks in the United Kingdom in 2018 Using a Pond-Side Test.
A Seasonal Study of Koi Herpesvirus and Koi Sleepy Disease Outbreaks in the United Kingdom in 2018 Using a Pond-Side Test.
Animals : an open access journal from MDPI (2021-02-13)
Irene Cano, John Worswick, Brian Mulhearn, David Stone, Gareth Wood, Jacqueline Savage, Richard Paley
ABSTRACT
Fluorescence real-time LAMP assays were designed for the orf43 gene of CyHV-3 European genotype and the p4a gene of the CEV genogroup I. A third LAMP assay to detect the ef1a gene of the host common carp was designed as an internal control. The limit of detection was 102 and 103 viral copies under 25 min for CyHV-3 and CEV, respectively. The specificity of the CyHV-3 LAMP assay was 95.6% of 72 fish herpesviruses tested. Sixty-three non-lethal common carp mucus swabs were collected across 16 sites during disease investigations. DNA extractions were performed in under 10 min using the QuickExtract™ digestion buffer. The LAMP amplification of CyHV-3 DNA in mucus swabs from clinical cases was detected from 4 to 13 min in 13 sites, while a co-infection of CyHV-3 and CEV was confirmed by LAMP in a single site. The LAMP results agreed with the results of the reference laboratory. The common carp ef1a was amplified only in 61% of the mucus swabs collected, preventing its use as a robust internal control to distinguish false negatives from invalid tests. After further optimization, these tests could be implemented for border inspection posts surveillance and decentralizing testing during disease outbreaks.
Product Description
CCB cell line from carp, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.