Merck
  • IGF1 receptor inhibition amplifies the effects of cancer drugs by autophagy and immune-dependent mechanisms.
IGF1 receptor inhibition amplifies the effects of cancer drugs by autophagy and immune-dependent mechanisms.
Journal for immunotherapy of cancer (2021-06-16)
Qi Wu, Ai-Ling Tian, Bei Li, Marion Leduc, Sabrina Forveille, Peter Hamley, Warren Galloway, Wei Xie, Peng Liu, Liwei Zhao, Shuai Zhang, Pan Hui, Frank Madeo, Yi Tu, Oliver Kepp, Guido Kroemer
ABSTRACT
Pharmacological autophagy enhancement constitutes a preclinically validated strategy for preventing or treating most major age-associated diseases. Driven by this consideration, we performed a high-content/high-throughput screen on 65 000 distinct compounds on a robotized fluorescence microscopy platform to identify novel autophagy inducers. Here, we report the discovery of picropodophyllin (PPP) as a potent inducer of autophagic flux that acts on-target, as an inhibitor of the tyrosine kinase activity of the insulin-like growth factor-1 receptor (IGF1R). Thus, PPP lost its autophagy-stimulatory activity in cells engineered to lack IGF1R or to express a constitutively active AKT serine/threonine kinase 1 (AKT1) mutant. When administered to cancer-bearing mice, PPP improved the therapeutic efficacy of chemoimmunotherapy with a combination of immunogenic cytotoxicants and programmed cell death 1 (PDCD1, better known as PD-1) blockade. These PPP effects were lost when tumors were rendered PPP-insensitive or autophagy-incompetent. In combination with chemotherapy, PPP enhanced the infiltration of tumors by cytotoxic T lymphocytes, while reducing regulatory T cells. In human triple-negative breast cancer patients, the activating phosphorylation of IGF1R correlated with inhibited autophagy, an unfavorable local immune profile, and poor prognosis. Altogether, these results suggest that IGF1R may constitute a novel and druggable therapeutic target for the treatment of cancer in conjunction with chemoimmunotherapies.
Product Description
Roche
Anti-HA High Affinity, from rat IgG1
Pricing and availability is not currently available.