Merck
Biological Monitoring of Metal Ions Released from Hip Prostheses.
International journal of environmental research and public health (2020-05-10)
Annamaria Nicolli, Andrea Trevisan, Isabella Bortoletti, Assunta Pozzuoli, Pietro Ruggieri, Andrea Martinelli, Alberto Gambalunga, Mariella Carrieri
ABSTRACT
The aim of this study was to evaluate the levels of As, Be, Bi, Cd, Co, Cr, Cu, Hg, Mn, Ni, Pb, Se, Tl, V, and Zn, by inductively coupled plasma-mass spectrometry (ICP-MS) in the urine of two groups of patients with two different types of metal-on-metal (MoM) total hip prostheses (ASR DePuy®, group A, 25 patients; total Met-Met System Lima®, group B, 28 patients). The determination of metals reflected a steady-state release (group A: 9 years after surgery and group B: 6 years after surgery). The results obtained confirmed the increase of Co and Cr urinary levels in both group when compared with the reference values for the general population adopted by the Italian Society of Reference Values (SIVR). In particular, Co and Cr levels exceeded the threshold values in urine, respectively, of 30 μg and 21 µg, adjusted to creatinine based on the threshold in whole blood of 7 μg/L proposed by the Medicines and Healthcare Products Regulatory Agency (MHRA). Regarding the other investigated metals, significantly higher values were found in Group A than in Group B. These differences could be due to the type of hip prosthesis implanted, the longer period of time since the implantation, as well as many other factors such as diet, age, drug consumption, physical activity, or presence of dental fillings. The continuous monitoring over the years of metal concentrations in patients carrying a prosthesis could be useful to better identify the sources of these metals.
Product Description
Milli-Q
Milli-Q® IQ 7010 Ultrapure and Pure Water Purification System, Produces ultrapure (Type 1) water and pure (Type 2) water with a production flow rate of 10 L/hr from tap water feed.
Milli-Q
Milli-Q® IQ 7003/05/10/15 Water Purification Systems, Produces ultrapure (Type 1) water and pure (Type 2) water with a production flow rate of 5 L/hr from tap water feed.
Milli-Q
Milli-Q® IQ 7003/05/10/15 Water Purification Systems, Produces ultrapure (Type 1) water and pure (Type 2) water with a production flow rate of 15 L/hr from tap water feed.
Milli-Q
Milli-Q® IQ 7003/05/10/15 Water Purification Systems, Produces ultrapure (Type 1) water and pure (Type 2) water with a production flow rate of 3 L/hr from tap water feed