Merck
  • The Mammalian Cap-Specific m6Am RNA Methyltransferase PCIF1 Regulates Transcript Levels in Mouse Tissues.
The Mammalian Cap-Specific m6Am RNA Methyltransferase PCIF1 Regulates Transcript Levels in Mouse Tissues.
Cell reports (2020-08-20)
Radha Raman Pandey, Elena Delfino, David Homolka, Adriana Roithova, Kuan-Ming Chen, Lingyun Li, Giulia Franco, Cathrine Broberg Vågbø, Emmanuel Taillebourg, Marie-Odile Fauvarque, Ramesh S Pillai
ABSTRACT
The 5' end of eukaryotic mRNAs is protected by the m7G-cap structure. The transcription start site nucleotide is ribose methylated (Nm) in many eukaryotes, whereas an adenosine at this position is further methylated at the N6 position (m6A) by the mammalian Phosphorylated C-terminal domain (CTD)-interacting Factor 1 (PCIF1) to generate m6Am. Here, we show that although the loss of cap-specific m6Am in mice does not affect viability or fertility, the Pcif1 mutants display reduced body weight. Transcriptome analyses of mutant mouse tissues support a role for the cap-specific m6Am modification in stabilizing transcripts. In contrast, the Drosophila Pcif1 is catalytically dead, but like its mammalian counterpart, it retains the ability to associate with the Ser5-phosphorylated CTD of RNA polymerase II (RNA Pol II). Finally, we show that the Trypanosoma Pcif1 is an m6Am methylase that contributes to the N6,N6,2'-O-trimethyladenosine (m62Am) in the hypermethylated cap4 structure of trypanosomatids. Thus, PCIF1 has evolved to function in catalytic and non-catalytic roles.
Sigma-Aldrich
Bouin′s solution, histological fixative
Sigma-Aldrich
Anti-PCIF1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Ponceau S, BioReagent, suitable for electrophoresis