Merck
  • B Cell, Th17, and Neutrophil Related Cerebrospinal Fluid Cytokine/Chemokines Are Elevated in MOG Antibody Associated Demyelination.
B Cell, Th17, and Neutrophil Related Cerebrospinal Fluid Cytokine/Chemokines Are Elevated in MOG Antibody Associated Demyelination.
PloS one (2016-02-27)
Kavitha Kothur, Louise Wienholt, Esther M Tantsis, John Earl, Sushil Bandodkar, Kristina Prelog, Fiona Tea, Sudarshini Ramanathan, Fabienne Brilot, Russell C Dale
ABSTRACT
Myelin oligodendrocyte glycoprotein antibody (MOG Ab) associated demyelination represents a subgroup of autoimmune demyelination that is separate from multiple sclerosis and aquaporin 4 IgG-positive NMO, and can have a relapsing course. Unlike NMO and MS, there is a paucity of literature on immunopathology and CSF cytokine/chemokines in MOG Ab associated demyelination. To study the differences in immunopathogenesis based on cytokine/chemokine profile in MOG Ab-positive (POS) and -negative (NEG) groups. We measured 34 cytokines/chemokines using multiplex immunoassay in CSF collected from paediatric patients with serum MOG Ab POS [acute disseminated encephalomyelitis (ADEM = 8), transverse myelitis (TM = 2) n = 10] and serum MOG Ab NEG (ADEM = 5, TM = 4, n = 9) demyelination. We generated normative data using CSF from 20 non-inflammatory neurological controls. The CSF cytokine and chemokine levels were higher in both MOG Ab POS and MOG Ab NEG demyelination groups compared to controls. The CSF in MOG Ab POS patients showed predominant elevation of B cell related cytokines/chemokines (CXCL13, APRIL, BAFF and CCL19) as well as some of Th17 related cytokines (IL-6 AND G-CSF) compared to MOG Ab NEG group (all p<0.01). In addition, patients with elevated CSF MOG antibodies had higher CSF CXCL13, CXCL12, CCL19, IL-17A and G-CSF than patients without CSF MOG antibodies. Our findings suggest that MOG Ab POS patients have a more pronounced CNS inflammatory response with elevation of predominant humoral associated cytokines/chemokines, as well as some Th 17 and neutrophil related cytokines/chemokines suggesting a differential inflammatory pathogenesis associated with MOG antibody seropositivity. This cytokine/chemokine profiling provides new insight into disease pathogenesis, and improves our ability to monitor inflammation and response to treatment. In addition, some of these molecules may represent potential immunomodulatory targets.
Product Description
Millipore
MILLIPLEX® Human Cytokine/Chemokine Magnetic Bead Panel III - Immunology Multiplex Assay, Simultaneously analyze multiple cytokine and chemokine biomarkers with Bead-Based Multiplex Assays using the Luminex technology, in mouse serum, plasma and cell culture samples.
Millipore
MILLIPLEX® Human Cytokine/Chemokine Magnetic Bead Panel II - Immunology Multiplex Assay, Simultaneously analyze multiple cytokine and chemokine biomarkers with Bead-Based Multiplex Assays using the Luminex technology, in human serum, plasma and cell culture samples.