Merck
  • Influence of influenza A infection on capsaicin-induced responses in murine airways.
Influence of influenza A infection on capsaicin-induced responses in murine airways.
The Journal of pharmacology and experimental therapeutics (2011-11-09)
Samuel J Taylor, Tracy S Mann, Peter J Henry
ABSTRACT
The principal aim of the study was to determine the influence of influenza A virus infection on capsaicin-induced relaxation responses in mouse isolated tracheal segments and clarify the underlying mechanisms. Anesthetized mice were intranasally inoculated with influenza A/PR-8/34 virus (VIRUS) or vehicle (SHAM), and 4 days later tracheal segments were harvested for isometric tension recording and biochemical and histologic analyses. Capsaicin induced dose-dependent relaxation responses in carbachol-contracted SHAM trachea (e.g., 10 μM capsaicin produced 66 ± 4% relaxation; n = 11), which were significantly inhibited by capsazepine [transient receptor potential vanilloid type 1 (TRPV1) antagonist], (2S,3S)-3-{[3,5-bis(trifluoromethyl)phenyl]methoxy}-2-phenylpiperidine hydrochloride (L-733,060) [neurokinin 1 (NK₁) receptor antagonist], indomethacin [cyclooxygenase (COX) inhibitor], and the combination of 6-isopropoxy-9-oxoxanthene-2-carboxylic acid (AH6809) and 7-[5α-([1S,1α(Z)-biphenyl]-4-ylmethoxy)-2β-(4-morpholinyl)-3-oxocyclopentyl]-4-heptenoic acid, calcium salt, hydrate (AH23848) [E-prostanoid (EP)₂ and EP₄ receptor antagonists, respectively], indicating that capsaicin-induced relaxation involved the TRPV1-mediated release of substance P (SP), activation of epithelial NK₁ receptors, and production of COX products capable of activating relaxant EP₂/EP₄ receptors. Consistent with this postulate, capsaicin-induced relaxation was associated with the significant release of SP and prostaglandin E₂ (PGE₂) from mouse tracheal segments. As expected, influenza A virus infection was associated with widespread disruption of the tracheal epithelium. Tracheal segments from VIRUS mice responded weakly to capsaicin (7 ± 3% relaxation) and were 25-fold less responsive to SP than tracheas from SHAM mice. In contrast, relaxation responses to exogenous PGE₂ and the β-adrenoceptor agonist isoprenaline were not inhibited in VIRUS trachea. Virus infection was associated with impaired capsaicin-induced release of PGE₂, but the release of SP was not affected. In summary, influenza A virus infection profoundly inhibits capsaicin- and SP-induced relaxation responses, most likely by inhibiting the production of PGE₂.
Sigma-Aldrich
AH6809, ≥98%, crystalline solid or supercooled liquid
Sigma-Aldrich
AH23848 hemicalcium salt, ≥90% (HPLC), powder