Merck
  • Impact factors on the production of β-methylamino-L-alanine (BMAA) by cyanobacteria.
Impact factors on the production of β-methylamino-L-alanine (BMAA) by cyanobacteria.
Chemosphere (2019-11-24)
Boyin Yan, Zhiquan Liu, Rui Huang, Yongpeng Xu, Dongmei Liu, Wei Wang, Zhiwei Zhao, Fuyi Cui, Wenxin Shi
ABSTRACT
Cyanobacteria produce a series of secondary metabolites, one of which is beta-N-methylamino-l-alanine (BMAA). BMAA is considered to be the cause of human neurodegeneration. Compared with other cyanotoxins, the role of BMAA in cyanobacteria remains unclear. To investigate this question, six strains of cyanobacteria were cultured and tested in this experiment with an optimized and validated BMAA determination method. The results show that four strains can produce BMAA. The effects of nutrient levels on the production of BMAA by Anabaena sp. FACHB-418 were studied by changing the initial concentrations of nitrate (NaNO3) and phosphate (K2HPO4) in mediums. Bound BMAA was detected in all samples and the concentrations were within 50-100 ng/g. Free BMAA was presence when the concentration of nitrogen was lower than 1.7 mg/L (121.43 μM). Free BMAA was released from the dead and ruptured cells during the cell decline period, so dissolved BMAA cannot be detectable in the adaptation and logarithmic periods, but could be abundant in the decline periods. Statistical analyses show that free BMAA concentrations were negatively correlated with nitrogen strongly (p = 2.334 × 10-10 and r = -0.842), but positively correlated with phosphorus weakly (p = 0.016 and r = 0.405). Moreover, the results of culture experiments indicated that exogenous BMAA could inhibit the growth of cyanobacteria that cannot produce BMAA, and the effect was enhanced as the concentration of exogenous BMAA increased. This phenomenon implies that the production of BMAA may be the stress response by some cyanobacteria to low nitrogen conditions to kill other cyanobacteria, i.e., their competitors.
Sigma-Aldrich
L-BMAA hydrochloride, ≥97% (NMR), powder