Merck
  • Toxoplasma gondii TgIST co-opts host chromatin repressors dampening STAT1-dependent gene regulation and IFN-γ-mediated host defenses.
Toxoplasma gondii TgIST co-opts host chromatin repressors dampening STAT1-dependent gene regulation and IFN-γ-mediated host defenses.
The Journal of experimental medicine (2016-08-10)
Gabrielle Gay, Laurence Braun, Marie-Pierre Brenier-Pinchart, Julien Vollaire, Véronique Josserand, Rose-Laurence Bertini, Aurélie Varesano, Bastien Touquet, Pieter-Jan De Bock, Yohann Coute, Isabelle Tardieux, Alexandre Bougdour, Mohamed-Ali Hakimi
ABSTRACT
An early hallmark of Toxoplasma gondii infection is the rapid control of the parasite population by a potent multifaceted innate immune response that engages resident and homing immune cells along with pro- and counter-inflammatory cytokines. In this context, IFN-γ activates a variety of T. gondii-targeting activities in immune and nonimmune cells but can also contribute to host immune pathology. T. gondii has evolved mechanisms to timely counteract the host IFN-γ defenses by interfering with the transcription of IFN-γ-stimulated genes. We now have identified TgIST (T. gondii inhibitor of STAT1 transcriptional activity) as a critical molecular switch that is secreted by intracellular parasites and traffics to the host cell nucleus where it inhibits STAT1-dependent proinflammatory gene expression. We show that TgIST not only sequesters STAT1 on dedicated loci but also promotes shaping of a nonpermissive chromatin through its capacity to recruit the nucleosome remodeling deacetylase (NuRD) transcriptional repressor. We found that during mice acute infection, TgIST-deficient parasites are rapidly eliminated by the homing Gr1(+) inflammatory monocytes, thus highlighting the protective role of TgIST against IFN-γ-mediated killing. By uncovering TgIST functions, this study brings novel evidence on how T. gondii has devised a molecular weapon of choice to take control over a ubiquitous immune gene expression mechanism in metazoans, as a way to promote long-term parasitism.
Sigma-Aldrich
ChIPAb+ Acetyl-Histone H3 (Lys27) - ChIP Validated Antibody and Primer Set, clone CMA309, from mouse
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-acetyl-Histone H4 Antibody, serum, Upstate®
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
ChIPAb+ Acetyl-Histone H3 (Lys9) Purified - ChIP Validated Antibody and Primer Set, from rabbit, purified by using Protein A