Merck
  • Heterotrimeric kinesin-2, together with kinesin-1, steers vesicular acetylcholinesterase movements toward the synapse.
Heterotrimeric kinesin-2, together with kinesin-1, steers vesicular acetylcholinesterase movements toward the synapse.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2016-12-07)
Anuttama Kulkarni, Yasmin Khan, Krishanu Ray
ABSTRACT
Acetylcholinesterase (AChE), which is implicated in the pathophysiology of neurological disorders, is distributed along the axon and enriched at the presynaptic basal lamina. It hydrolyses the neurotransmitter acetylcholine, which inhibits synaptic transmission. Aberrant AChE activity and ectopic axonal accumulation of the enzyme are associated with neurodegenerative disorders, such as Alzheimer's disease. The molecular mechanism that underlies AChE transport is still unclear. Here, we show that expression of Drosophila AChE tagged with photoactivatable green fluorescent protein and m-Cherry (GPAC) in cholinergic neurons compensates for the RNA interference-mediated knockdown of endogenous AChE activity. GPAC-AChE, which is enriched in the neuropil region of the brain, moves in the apparently vesicular form in axons with an anterograde bias in Drosophila larvae. Two anterograde motors, kinesin-1 and -2, propel distinct aspects of GPAC-AChE movements. Total loss of kinesin-2 reduces the density of anterograde traffic and increases bidirectional movements of GPAC-AChE vesicles without altering their speed. A partial loss of kinesin-1 reduces both the density and speed of anterograde GPAC-AChE traffic and enhances the pool of stationary vesicles. Together, these results suggest that combining activity of a relatively weak kinesin-2 with that of a stronger kinesin-1 motor could steer AChE-containing vesicles toward synapse, and provides a molecular basis for the observed subcellular distribution of the enzyme.-Kulkarni, A., Khan, Y., Ray, K. Heterotrimeric kinesin-2, together with kinesin-1, steers vesicular acetylcholinesterase movements toward the synapse.
Sigma-Aldrich
Anti-Synaptotagmin antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-ACHE antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Kinesin Antibody, heavy chain, a.a.420-445, clone H2, clone H2, Chemicon®, from mouse
Sigma-Aldrich
Anti-Kinesin Antibody, light chain, clone L2, clone L2, Chemicon®, from mouse
Sigma-Aldrich
Anti-KIF3A antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution