Merck
  • PTPN3 acts as a tumor suppressor and boosts TGF-β signaling independent of its phosphatase activity.
PTPN3 acts as a tumor suppressor and boosts TGF-β signaling independent of its phosphatase activity.
The EMBO journal (2019-07-16)
Bo Yuan, Jinquan Liu, Jin Cao, Yi Yu, Hanchenxi Zhang, Fei Wang, Yezhang Zhu, Mu Xiao, Sisi Liu, Youqiong Ye, Le Ma, Dewei Xu, Ningyi Xu, Yi Li, Bin Zhao, Pinglong Xu, Jianping Jin, Jianming Xu, Xi Chen, Li Shen, Xia Lin, Xin-Hua Feng
ABSTRACT
TGF-β controls a variety of cellular functions during development. Abnormal TGF-β responses are commonly found in human diseases such as cancer, suggesting that TGF-β signaling must be tightly regulated. Here, we report that protein tyrosine phosphatase non-receptor 3 (PTPN3) profoundly potentiates TGF-β signaling independent of its phosphatase activity. PTPN3 stabilizes TGF-β type I receptor (TβRI) through attenuating the interaction between Smurf2 and TβRI. Consequently, PTPN3 facilitates TGF-β-induced R-Smad phosphorylation, transcriptional responses, and subsequent physiological responses. Importantly, the leucine-to-arginine substitution at amino acid residue 232 (L232R) of PTPN3, a frequent mutation found in intrahepatic cholangiocarcinoma (ICC), disables its role in enhancing TGF-β signaling and abolishes its tumor-suppressive function. Our findings have revealed a vital role of PTPN3 in regulating TGF-β signaling during normal physiology and pathogenesis.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
IgG from mouse serum, reagent grade, ≥95% (SDS-PAGE), lyophilized powder
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
SB 431542 hydrate, ≥98% (HPLC), powder