Merck
  • Nicotinamide phosphoribosyltransferase is a molecular target of potent anticancer agents identified from phenotype-based drug screening.
Nicotinamide phosphoribosyltransferase is a molecular target of potent anticancer agents identified from phenotype-based drug screening.
Scientific reports (2019-05-28)
Daisuke Yamaguchi, Takamichi Imaizumi, Kaori Yagi, Yuichi Matsumoto, Takayuki Nakashima, Akiyo Hirose, Naomi Kashima, Yukino Nosaka, Tomoko Hamada, Katsuya Okawa, Yoichi Nishiya, Kazuo Kubo
ABSTRACT
Phenotypic screening in drug discovery has been revived with the expectation of providing promising lead compounds and drug targets and improving the success rate of drug approval. However, target identification remains a major bottleneck in phenotype-based drug discovery. We identified the lead compounds K542 and K405 with a selective inhibition of cell viability against sphingosine-1-phosphate lyase 1 (SGPL1)-transduced ES-2 cells by phenotypic screening. We therefore performed an in vivo pharmacological examination and observed the antitumor activity of K542 in an HT-1080 tumor-bearing mouse xenograft model. SGPL1 was expected to be a therapeutic target in some cancers, suggesting that these lead molecules might be promising candidates; however, their mechanisms of action still remain unexplained. We therefore synthesized the affinity probe Ind-tag derived from K542 and identified the proteins binding to Ind-tag via a pull-down experiment. Proteomics and biochemical analyses revealed that the target molecule of these lead compounds was Nicotinamide phosphoribosyltransferase (NAMPT). We established K542-resistant DLD-1 and HT-1080 cells, and genetic analyses of these cells identified a missense mutation in the NAMPT-encoding gene. This enzymatic experiment clearly showed that K393 exerts enzymatic inhibition against NAMPT. These proteomics, genetics and biochemical analyses clarified that compounds K542 and K405 were NAMPT inhibitors.
Sigma-Aldrich
Paclitaxel, from Taxus brevifolia, ≥95% (HPLC), powder